Airfree® Schlenk Hose Adapters, Chemglass
Supplier: Chemglass
Certificates
About this item
Adapters for use with manifolds such as (80060-120) through (80060-128) to permit port connection to standard rubber hose.
- Adapters have hooks just below the grind
- For use with (80072-944) stainless steel springs
Adapters have hooks just below the grind for use with (80072-944) stainless steel springs. The 24/40 size is available with a constriction above the serrated hose connection to permit bowl area to be filled with glass wool, preventing solids from being drawn into the manifold. Size of hose connection is at the middle serration.