


Ammonia solution 28 - 30% ACS
Catalog # 10119-806
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:10119-806
- Supplier no.:325690
Specifications
Specification Test Results
- Appearance:Colorless, free from suspended matter
- Assay (w/w):28.0 - 30.0%
- Sulfate :2 ppm maximum
- Residue after Ignition:20 ppm maximum
- Carbon Dioxide:20 ppm maximum
- Chloride:0.5 ppm maximum
- Nitrate:2 ppm maximum
- Phosphate:2 ppm maximum
- Reducing Substances Permanganate:Passes Test
- Heavy Metals:0.5 ppm maximum
- Iron:0.2 ppm maximum
Specification Test Results
About this item
:
:35.05 g/mol
UN:2672
ADR:8,III
:GHS05
:GHS07
:GHS09
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650