Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TNF Rabbit Monoclonal Antibody [clone: D2E7 (Adalimumab)]
Anti-TNF Rabbit Monoclonal Antibody [clone: D2E7 (Adalimumab)]
  76284-582
Anti-TNF Rabbit Monoclonal Antibody [clone: D2E7 (Adalimumab)]
  76284-582
 :  ABS565-0200
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    TNF-alpha
  • Antigen Symbol:
    TNF
  • Clonality:
    Monoclonal
  • Clone:
    D2E7 (Adalimumab)
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Size:
    200 µg
  • Form:
    Liquid
  • Antigen Synonyms:
    Tumor necrosis factor ligand superfamily member 2|Cachectin|Tumor necrosis factor-α|TNFSF 2
  • UniProtKB:
    UniProt# P01375
  • Storage Buffer:
    In PBS containing 0.02% Proclin 300.
  • Storage Temperature:
    –20 °C
  • Shipping Temperature:
    Shipped on Blue Ice
  • Immunogen:
    This antibody was generated by affinity maturation from the 2SD4 scFv, which in turn was selected on human TNF alpha, using the murine anti-hTNF alpha antibody Mab32 as a template.
  • Purification:
    Protein A affinity purified.
  • Cat. No.:
    76284-582

 

 

Recombinant monclonal antibody (Rabbit IgGκ).

Recommended Applications: ELISA, IF, IHC, FUNC

Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: D2E7 (Adalimumab)
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human