Order Entry
United States
ContactUsLinkComponent
Anti-CD19 Mouse Monoclonal Antibody [clone: FMC63]
Anti-CD19 Mouse Monoclonal Antibody [clone: FMC63]
Catalog # 76284-622
Supplier:  Enzo Life Sciences
Anti-CD19 Mouse Monoclonal Antibody [clone: FMC63]
Catalog # 76284-622
Supplier:  Enzo Life Sciences
Supplier Number:  ABS415-0200
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD19
  • Antigen Symbol:
    CD19
  • Clonality:
    Monoclonal
  • Clone:
    FMC63
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Size:
    200 µg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen Synonyms:
    B-lymphocyte antigen CD19|Differentiation antigen CD19 T-cell surface antigen Leu-12|B-lymphocyte surface antigen B4
  • UniProtKB:
    UniProt# P15391
  • Storage Buffer:
    In PBS containing 0.02% Proclin 300.
  • Storage Temperature:
    –20 °C
  • Shipping Temperature:
    Shipped on Blue Ice
  • Immunogen:
    Human prolymphocytic leukaemia cell line JVM3.
  • Purification:
    Protein A affinity purified.
  • Cat. No.:
    76284-622

Specifications

About this item

Recombinant monclonal antibody (Mouse IgG2aκ).

Recommended Applications: Flow Cytometry, IF

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: FMC63
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: Human