ApoScreen® Human Recombinant Annexin V (from E. coli), FITC (Fluorescein Isothiocyanate)
Catalog # 103265-462
Supplier: Southern Biotechnology
CAS Number:
Specifications
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 tests
- Storage Conditions:2-8 °C
- Gene ID:NCBI 308
- Protein Synonyms:ANX5|AXV|Annexin A5
- Protein/Peptide Name:Annexin V
- Concentration:Lot specific
- Formulation:Supplied in ApoScreen® Annexin V Binding Buffer
- Shipping Temperature:Ambient
- Cat. No.:103265-462
- Supplier no.:10040-02
Specifications
About this item
ApoScreen® Annexin V (SB Cat. No. 10040) is a recombinant protein that is expressed in E. coli. A number of changes in cell structure occur during apoptosis. One major change is the loss of membrane phospholipid asymmetry, with translocation of phosphatidylserine (PS) from the inner leaflet of the phospholipid bilayer to the external surface. Annexin V, a member of the annexin family of calcium-dependent phospholipid binding proteins, has a high affinity for phosphatidylserine (PS)-containing phospholipid bilayers. Conjugated Annexin V can be used to monitor changes in cell membrane phospholipid asymmetry, thereby providing a convenient tool for detection of apoptotic cells.