ApoScreen® Human Recombinant Annexin V (from E. coli)
103265-460
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.1 mg
- Storage Conditions:–20 °C prior to reconstitution; 2-8 °C following reconstitution
- Gene ID:NCBI 308
- Reconstitution Instructions:Reconstitute with 1.0 mL of dH2O
- Protein Synonyms:ANX5|AXV|Annexin A5
- Protein/Peptide Name:Annexin V
- Formulation:Supplied lyophilized from phosphate buffered saline
- Shipping Temperature:Ambient
- Cat. No.:103265-460
- Supplier no.:10040-01
ApoScreen® Annexin V (SB Cat. No. 10040) is a recombinant protein that is expressed in E. coli. A number of changes in cell structure occur during apoptosis. One major change is the loss of membrane phospholipid asymmetry, with translocation of phosphatidylserine (PS) from the inner leaflet of the phospholipid bilayer to the external surface. Annexin V, a member of the annexin family of calcium-dependent phospholipid binding proteins, has a high affinity for phosphatidylserine (PS)-containing phospholipid bilayers. Conjugated Annexin V can be used to monitor changes in cell membrane phospholipid asymmetry, thereby providing a convenient tool for detection of apoptotic cells.