Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Mouse Kappa Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 187.1]
Anti-Mouse Kappa Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 187.1]
Certificates
Anti-Mouse Kappa Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 187.1]

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • 100242-854
  • 100242-856
  • Antigen Symbol:
  • Kappa
  • Kappa
  • Antigen Name:
  • Kappa
  • Kappa
  • Antibody Type:
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 187.1
  • 187.1
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 243469
  • 243469
  • Isotype:
  • IgG1
  • IgG1
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • MPC-11 immunoglobulin
  • MPC-11 immunoglobulin
  • Form:
  • Liquid
  • Liquid
  • Concentration:
  • 0.5 mg/mL
  • 0.5 mg/mL
  • Storage Buffer:
  • Supplied in PBS/NaN3
  • Supplied in PBS/NaN3
  • Storage Temperature:
  • 2-8°C
  • 2-8°C
  • Shipping Temperature:
  • Ambient Temperature
  • Ambient Temperature
  • Tested Applications:
  • FLISA
  • FLISA

Specifications