Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Control
- Antigen Name:Keyhole limpet hemocyanin
- Antigen Symbol:KLH
- Clonality:Monoclonal
- Clone:KLH/G2a-1-1
- Conjugation:Cy5®
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Size:0.1 mg
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Tested Applications:FLISA
- Cat. No.:100242-096
- Supplier no.:0117-15
Specifications
About this item
Rat IgG2a-CY5 for use as a control in flow cytometry and immunohistochemistry assays.
Type: Control
Antigen: KLH
Clonality: Monoclonal
Clone: KLH/G2a-1-1
Conjugation: Cy5®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: