Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Control
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:10 mg
- ChIP:Yes
- Form:Liquid
- Storage Buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
- Storage Temperature:2-8°C
- Concentration:5 mg/mL
- Shipping Temperature:Ambient Temperature
- Tested Applications:Microarray, Block, In vitro Control
- Cat. No.:100242-038
- Supplier no.:0111-01
Specifications
About this item
Purified Rabbit IgG for use as a control in flow cytometry, immunocytochemistry / immunocytochemistry, western blotting, immunoprecipitation, ChIP, microarray, and in vitro assays and as a standard in ELISA.
Type: Control
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: