Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Control
- Antigen Name:Immunoglobulin A
- Antigen Symbol:IgA
- Clonality:Monoclonal
- Clone:A-1
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Chicken
- Size:100 tests
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Tested Applications:FLISA
- Cat. No.:100241-940
- Supplier no.:0104-11
Specifications
About this item
Mouse IgG2b-APC for use as a control in flow cytometry assays.
Type: Control
Antigen: IgA
Clonality: Monoclonal
Clone: A-1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Chicken