ContactUsLinkComponent
CellTrics® Disposable Cell Strainers, Sysmex Amercica
CellTrics® Disposable Cell Strainers, Sysmex Amercica
Catalog # MSPP-40042327
CellTrics® Disposable Cell Strainers, Sysmex Amercica
Catalog # MSPP-40042327
Supplier Number:  04-004-2327
Specifications
  • Pore size:
    50 μm
  • Sterility:
    Sterile
  • Color:
    Yellow
  • Description:
    CellTrics Cell Strainers
  • Cat. No.:
    MSPP-40042327
Specifications
About this item
CellTrics® are disposable filters for isolation of cells and nuclei from cell debris and aggregates.
  • Not autoclavable
  • One-time use only
  • No additional devices are required
  • 5 µm mesh diameter
These sterile filters are single-packed and have tilted filters made from monofil nylon material.
Caution: For research use only.