Neutralizing Buffer Swabs, GVS Filter Technology
Supplier: GVS FILTER TECHNOLOGY
Certificates
About this item
Neutralizing buffer swabs are used in the monitoring of surfaces for total bacterial count.
Neutralizing buffer inactivates the bactericidal and bacteriostatic effects of chlorine and quaternary ammonium detergents. Without exhibiting no toxic effects on microorganisms, this permits the transfer of swabbed organisms to the laboratory without loss in viability.
Caution: Not designed to culture and enumerate microorganisms.