Digital Thermometers, Triple Display, Chemglass
The triple display digital thermometer simultaneously shows the current, minimum, and maximum temperatures and updates continuously. An audible alarm will sound when the temperature rises above or goes below the set temperatures.
- Temperature probe housed in plastic bottle filled with bio-safe glycol water mixture
- Instrument is ºC and ºF switchable
- Thermometer can be used to monitor ambient temperatures
- Audible alarm with one critical temperature
The minimum or maximum feature monitors critical temperature for any period of time, such as overnight or weekend.
The thermometer probe is housed in an unbreakable plastic bottle or vial filled with a buffering media which maintains the thermometer reading when the instrument door is opened. The thermometer and the bottle housing the probe have dual magnets allowing attachment to doors or walls of the instrument.
: NIST Traceable, each thermometer individually serialized.
: Supplied with AAA alkaline battery, 9' long micro cable, flip-open stand and a wall mounting key hole.