Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant MMP-12
Human Recombinant MMP-12
Catalog # 220025-872
Supplier:  R&D Systems
CAS Number:  
Human Recombinant MMP-12
Catalog # 220025-872
Supplier:  R&D Systems
Supplier Number:  WBC019
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Size:
    1 Vial
  • Storage Conditions:
    Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
  • Gene ID:
    4321
  • Protein Synonyms:
    Macrophage elastase|hME|MGC138506|MMP-12|macrophage metalloelastase|Matrix metalloproteinase-12|MME|ME|EC 3.4.24|matrix metalloproteinase 12 (macrophage elastase)|HMEEC 3.4.24.65|MMP12|matrix metallopeptidase 12 (macrophage elastase)
  • Protein/Peptide Name:
    MMP-12
  • Cat. No.:
    220025-872
  • Supplier no.:
    WBC019

Specifications

About this item

The Recombinant Human MMP-12 Western Blot Standard Protein has been validated for the following applications: Western Blot.