Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant USP25 (from Baculovirus (Sf21 Insect Cells))
Human Recombinant USP25 (from Baculovirus (Sf21 Insect Cells))
Catalog # 102880-850
Supplier:  R&D Systems
CAS Number:  
Human Recombinant USP25 (from Baculovirus (Sf21 Insect Cells))
Catalog # 102880-850
Supplier:  R&D Systems
Supplier Number:  E-546-050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Sf21 Insect cells)
  • Species:
    Human
  • Size:
    50 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
  • Gene ID:
    29761
  • Protein Synonyms:
    EC 3.1.2.15|ubiquitin specific peptidase 25|EC 3.4.19.12|Deubiquitinating enzyme 25|Ubiquitin thiolesterase 25|ubiquitin specific protease USP2510USP21ubiquitin specific protease 25|ubiquitin-specific processing protease 25|Ubiquitin-specific-processing protease 25|ubiquitin thioesterase 25|USP on chromosome 21|ubiquitin carboxyl-terminal hydrolase 25
  • Accession No.:
    Q9UHP3
  • Protein/Peptide Name:
    USP25
  • Purity:
    >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain
  • Cat. No.:
    102880-850
  • Supplier no.:
    E-546-050

Specifications

About this item

The Recombinant Human His6-USP25 Protein from R&D Systems, powered by R&D Systems, powered by Boston Biochem is derived from Sf 21 (baculovirus). The Recombinant Human His6-USP25 Protein has been validated for the following applications: Enzyme Activity.