Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 μg
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
- Gene ID:9099
- Protein Synonyms:EC 3.1.2.15|EC 3.4.19.12|41 kDa ubiquitin-specific protease|USP9|Ubiquitin thiolesterase 2|ubiquitin specific protease 2|Ubiquitin-specific-processing protease 2|Deubiquitinating enzyme 2|ubiquitin specific peptidase 2|ubiquitin thioesterase 2|UBP41ubiquitin carboxyl-terminal hydrolase 2|ubiquitin specific protease 12|UBP41|ubiquitin specific protease 9
- Accession No.:O75604
- Protein/Peptide Name:USP2
- Purity:>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain
- Cat. No.:102880-836
- Supplier no.:E-504-050
The Recombinant Human USP2 Catalytic Domain Protein from R&D Systems, powered by R&D Systems, powered by Boston Biochem is derived from E. coli. The Recombinant Human USP2 Catalytic Domain Protein has been validated for the following applications: Enzyme Activity.