Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD40 Ligand (from E. coli)
Human Recombinant CD40 Ligand (from E. coli)
Catalog # 102878-462
Supplier:  R&D Systems
CAS Number:  
Human Recombinant CD40 Ligand (from E. coli)
Catalog # 102878-462
Supplier:  R&D Systems
Supplier Number:  AFL617
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    1 mg
  • Environmentally Preferable:
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 3 months, 2 to 8 C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    959
  • Protein Synonyms:
    CD40 antigen ligand|TNFSF5IMD3|CD154 antigen|hCD40L|member 5 (hyper-IgM syndrome)|TRAP|CD40LIGM|CD40L|TRAPtumor necrosis factor (ligand) superfamily|gp39|T-B cell-activating molecule|CD154|tumor necrosis factor (ligand) superfamily member 5|CD40-L|Tumor necrosis factor ligand superfamily member 5|CD40LG|TNF-related activation protein|T-BAM|TNFSF5|HIGM1|CD40 ligand|T-cell antigen Gp39
  • Accession No.:
    P29965
  • Protein/Peptide Name:
    CD40 Ligand
  • Purity:
    >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight:
    18 kDa, reducing conditions
  • Cat. No.:
    102878-462
  • Supplier no.:
    AFL617

Specifications

About this item

The Recombinant Human CD40 Ligand/TNFSF5 (108-261), Animal-Free from R&D Systems is derived from E. coli. The Recombinant Human CD40 Ligand/TNFSF5 (108-261), Animal-Free has been validated for the following applications: Bioactivity.