To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Sf21 Insect cells)
- Species:Mouse
- Size:100 μg
- Environmentally Preferable:
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin Content:<1.0 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:3563
- Protein Synonyms:interleukin 3 receptor|IL-3RA|IL3R alpha|IL3RX|IL3RA|CD123 antigen|interleukin-3 receptor subunit alpha|alpha (low affinity)|IL-3R subunit alpha|IL3RY|CD123|IL-3 receptor subunit alpha|hIL3Ra|MGC34174|hIL-3Ra|IL3RAY|IL3R|IL-3R-alpha
- Accession No.:P26952
- Protein/Peptide Name:IL-3 R alpha/CD123
- Purity:>90%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular Weight:70-80 kDa, reducing conditions
- Cat. No.:220025-795
- Supplier no.:983-MR-100
The Recombinant Mouse IL-3 R alpha/CD123 Fc Chimera Protein from R&D Systems is derived from Sf 21 (baculovirus). The Recombinant Mouse IL-3 R alpha/CD123 Fc Chimera Protein has been validated for the following applications: Bioactivity.