To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:NS0 cells
- Species:Human
- Size:25 μg
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin Content:<1.0 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:6352
- Protein Synonyms:T-cell specific protein p288|D17S136Enormally T-expressed|EoCP|RANTES|SIS-delta|Small-inducible cytokine A5|T cell-specific protein P228|Eosinophil chemotactic cytokine|small inducible cytokine subfamily A (Cys-Cys)|and presumably secreted|chemokine (C-C motif) ligand 5|member 5|SISd|small inducible cytokine A5 (RANTES)|TCP228T-cell-specific protein RANTES
- Accession No.:P13501 (CCL5) & P78423 (CX3CL1 Mucin-like Stalk)
- Protein/Peptide Name:CCL5
- Purity:>90%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular Weight:85-105 kDa, reducing conditions
- Cat. No.:220025-789
- Supplier no.:978-RN-025/CF
The Recombinant Human CCL5/RANTES (Mucin Stalk Chimera) from R&D Systems is derived from NS0. The Recombinant Human CCL5/RANTES (Mucin Stalk Chimera) has been validated for the following applications: Bioactivity.