Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant GDF-15 (from HEK293 Cells)
Human Recombinant GDF-15 (from HEK293 Cells)
Catalog # 102877-328
Supplier:  R&D Systems
CAS Number:  
Human Recombinant GDF-15 (from HEK293 Cells)
Catalog # 102877-328
Supplier:  R&D Systems
Supplier Number:  8146-GD-010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    10 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    9518
  • Protein Synonyms:
    growth differentiation factor 15|PLABNRG-1|growth/differentiation factor 15|NAG-1NSAID-regulated gene 1 protein|PDFGDF-15|MIC1Prostate differentiation factor|MIC-1|PTGF-beta|NSAID (nonsteroidal inflammatory drug)-activated protein 1|Placental bone morphogenetic protein|PTGFBPTGF-beta|GDF15|Macrophage inhibitory cytokine 1|NAG-1|PDF|MIC-1NSAID-activated gene 1 protein|PLAB|Placental TGF-beta
  • Accession No.:
    Q99988
  • Protein/Peptide Name:
    GDF-15
  • Purity:
    >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
  • Molecular Weight:
    12-14 kDa, reducing conditions
  • Cat. No.:
    102877-328
  • Supplier no.:
    8146-GD-010

Specifications

About this item

The Recombinant Human GDF-15 (Human Cell-expressed) Protein from R&D Systems is derived from HEK293. The Recombinant Human GDF-15 (Human Cell-expressed) Protein has been validated for the following applications: Bioactivity.