Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Activin RIB/ALK-4 (from NS0 cells)
Human Recombinant Activin RIB/ALK-4 (from NS0 cells)
Catalog # 220025-665
Supplier:  R&D Systems
CAS Number:  
Human Recombinant Activin RIB/ALK-4 (from NS0 cells)
Catalog # 220025-665
Supplier:  R&D Systems
Supplier Number:  808-AR-100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    NS0 cells
  • Species:
    Human
  • Size:
    100 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    91
  • Protein Synonyms:
    SKR2activin receptor type-1B|Serine/threonine-protein kinase receptor R2|activin A receptor|ActRIB|Activin receptor-like kinase 4|ALK-4|ACVRLK4ACTR-IB|ALK4activin A receptor|EC 2.7.11.30|type II-like kinase 4|EC 2.7.11|type IB|ALK4|ActivinRIB|ACVR1B|Activin receptor type IB
  • Accession No.:
    AAA60556
  • Protein/Peptide Name:
    Activin RIB
  • Purity:
    >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Molecular Weight:
    45-50 kDa, reducing conditions
  • Cat. No.:
    220025-665
  • Supplier no.:
    808-AR-100

Specifications

About this item

The Recombinant Human Activin RIB/ALK-4 Fc Chimera Protein from R&D Systems is derived from NS0. The Recombinant Human Activin RIB/ALK-4 Fc Chimera Protein has been validated for the following applications: Binding Activity.