Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Aldo-keto Reductase 1C3/AKR1C3 (from E. coli)
Human Recombinant Aldo-keto Reductase 1C3/AKR1C3 (from E. coli)
Catalog # 102869-018
Supplier:  R&D Systems
CAS Number:  
Human Recombinant Aldo-keto Reductase 1C3/AKR1C3 (from E. coli)
Catalog # 102869-018
Supplier:  R&D Systems
Supplier Number:  7678-DH-020
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    20 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
  • Endotoxin Content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    8644
  • Protein Synonyms:
    aldo-keto reductase family 1|hydroxysteroid (17-beta) dehydrogenase 5|Testosterone 17-beta-dehydrogenase 5|type II)|member C3 (3-alpha hydroxysteroid dehydrogenase|AldoketoReductase 1C3|Dihydrodiol dehydrogenase type I|KIAA0119hluPGFS|EC 1.3.1.20|HAKRBtype IIb 3-alpha hydroxysteroid dehydrogenase|EC 1.1.1.188|DD317-beta-HSD 5|Trans-1|Dihydrodiol dehydrogenase 3|Indanol dehydrogenase|EC 1.1.1.64|DDH1|PGFS|DD3|HAKRe|2-dihydrobenzene-1|2-diol dehydrogenase|brain|EC 1.1.1.63|Prostaglandin F synthase|EC 1.1.1.213|aldo-keto reductase family 1 member C3|17-beta-hydroxysteroid dehydrogenase type 5|HAKRB|EC 1.1.1|3-alpha-HSD type 2,3-alpha-hydroxysteroid dehydrogenase type 2|EC 1.1.1.112,3-alpha-HSD type II|Chlordecone reductase homolog HAKRb|DDX|HSD17B5|HA1753|dihydrodiol dehydrogenase X|DD-3|AKR1C3
  • Accession No.:
    P42330
  • Protein/Peptide Name:
    Aldo-keto Reductase 1C3/AKR1C3
  • Purity:
    >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane
  • Molecular Weight:
    36-37 kDa, reducing conditions
  • Cat. No.:
    102869-018
  • Supplier no.:
    7678-DH-020

Specifications

About this item

The Recombinant Human Aldo-keto Reductase 1C3/AKR1C3 Protein from R&D Systems is derived from E. coli. The Recombinant Human Aldo-keto Reductase 1C3/AKR1C3 Protein has been validated for the following applications: Enzyme Activity.