Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant Caspase-3 (from E. coli)
Catalog # 220025-571
Supplier:  R&D Systems
CAS Number:  
Human Recombinant Caspase-3 (from E. coli)
Catalog # 220025-571
Supplier:  R&D Systems
Supplier Number:  731-C3-005
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    5 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Gene ID:
    836
  • Protein Synonyms:
    EC 3.4.22|apoptosis-related cysteine peptidase|proCaspase3|Yama|Protein Yama|Cysteine protease CPP32|Caspase3|CASP-3|PARP cleavage protease|CASP3|CPP32|CPP32SREBP cleavage activity 1|Apopain|apoptosis-related cysteine protease|EC 3.4.22.56|CPP32B|LICE-1|SCA-1|CPP-32|Caspase-3|Caspase 3
  • Accession No.:
    AAA65015
  • Protein/Peptide Name:
    Caspase-3
  • Purity:
    >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Molecular Weight:
    35 kDa, reducing conditions
  • Cat. No.:
    220025-571
  • Supplier no.:
    731-C3-005

Specifications

About this item

The Recombinant Human Pro-Caspase-3 Protein from R&D Systems is derived from E. coli. The Recombinant Human Pro-Caspase-3 Protein has been validated for the following applications: Enzyme Activity.