Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Aldo-keto Reductase 1C1/AKR1C1 (from E. coli)
Human Recombinant Aldo-keto Reductase 1C1/AKR1C1 (from E. coli)
Catalog # 102877-112
Supplier:  R&D Systems
CAS Number:  
Human Recombinant Aldo-keto Reductase 1C1/AKR1C1 (from E. coli)
Catalog # 102877-112
Supplier:  R&D Systems
Supplier Number:  6529-DH-020
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    20 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
  • Endotoxin Content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    1645
  • Protein Synonyms:
    aldo-keto reductase family 1|Chlordecone reductase homolog HAKRC|20-alpha-hydroxysteroid dehydrogenase|DDHH-37|EC 1.1.1.-|EC 1.3.1.20|EC 1.1.1.149|MBAB|EC 1.1.1.112|AKR1C1|High-affinity hepatic bile acid-binding protein|Trans-1|Indanol dehydrogenase|HAKRC|DDH1|2-ALPHA-HSD|HAKRCDDH1aldo-keto reductase C|20-alpha(3-alpha)-hydroxysteroid dehydrogenase)|2-dihydrobenzene-1|2-diol dehydrogenase|HBAB|20-ALPHA-HSD|EC 1.1.1|dihydrodiol dehydrogenase isoform DD1|aldo-keto reductase family 1 member C1|dihydrodiol dehydrogenase 1|Dihydrodiol dehydrogenase 1/2|member C1 (dihydrodiol dehydrogenase 1|hepatic dihydrodiol dehydrogenase|DD1/DD2|DD1MGC8954|type II 3-alpha-hydroxysteroid dehydrogenase|AldoketoReductase 1C1|C9|20 alpha-hydroxysteroid dehydrogenase
  • Accession No.:
    Q04828
  • Protein/Peptide Name:
    Aldo-keto Reductase 1C1/AKR1C1
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane.
  • Molecular Weight:
    36-38 kDa, reducing conditions
  • Cat. No.:
    102877-112
  • Supplier no.:
    6529-DH-020

Specifications

About this item

The Recombinant Human Aldo-keto Reductase 1C1/AKR1C1 Protein from R&D Systems is derived from E. coli. The Recombinant Human Aldo-keto Reductase 1C1/AKR1C1 Protein has been validated for the following applications: Enzyme Activity.