To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:10 μg
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin Content:<0.01 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:8600
- Protein Synonyms:tumor necrosis factor ligand superfamily member 11|Osteoprotegerin ligand|OPGLOPTB2|member 11|OPGL|sOdf|TNFSF11|CD254 antigen|ODF|tumor necrosis factor (ligand) superfamily|Receptor activator of nuclear factor kappa-B ligand|CD254|RANKLreceptor activator of nuclear factor kappa B ligand|TNF-related activation-induced cytokine|RANKL|Osteoclast differentiation factor|TRANCEODFhRANKL2|RANK L
- Accession No.:Q3TWY5
- Protein/Peptide Name:TRANCE/TNFSF11/RANK L
- Purity:>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
- Cat. No.:220024-780
- Supplier no.:462-TEC-010
The Recombinant Mouse TRANCE/RANK L/TNFSF11 (E. coli-expressed) from R&D Systems is derived from E. coli. The Recombinant Mouse TRANCE/RANK L/TNFSF11 (E. coli-expressed) has been validated for the following applications: Bioactivity.