Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant Sonic Hedgehog/Shh (from E. coli)
Mouse Recombinant Sonic Hedgehog/Shh (from E. coli)
Catalog # 220024-777
Supplier:  R&D Systems
CAS Number:  
Mouse Recombinant Sonic Hedgehog/Shh (from E. coli)
Catalog # 220024-777
Supplier:  R&D Systems
Supplier Number:  461-SH-025/CF
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    25 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    6469
  • Protein Synonyms:
    TPT|HLP3|sonic hedgehog|SMMCIsonic hedgehog homolog (Drosophila)|HHG1|sonic hedgehog protein|HPE3|Shh|sonic hedgehog homolog|TPTPS|HHG-1|MCOPCB5sonic hedgehog (Drosophila) homolog
  • Accession No.:
    Q62226
  • Protein/Peptide Name:
    Sonic Hedgehog/Shh
  • Purity:
    >97%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Cat. No.:
    220024-777
  • Supplier no.:
    461-SH-025/CF

Specifications

About this item

The Recombinant Mouse Sonic Hedgehog/Shh, N-Terminus Protein from R&D Systems is derived from E. coli. The Recombinant Mouse Sonic Hedgehog/Shh, N-Terminus Protein has been validated for the following applications: Bioactivity.