Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CD14 (from CHO cells)
Human Recombinant CD14 (from CHO cells)
Catalog # 220024-424
Supplier:  R&D Systems
CAS Number:  
Human Recombinant CD14 (from CHO cells)
Catalog # 220024-424
Supplier:  R&D Systems
Supplier Number:  383-CD-050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    50 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    929
  • Protein Synonyms:
    monocyte differentiation antigen CD14|CD14 antigen|CD14 molecule|CD14|Myeloid cell-specific leucine-rich glycoprotein
  • Accession No.:
    P08571
  • Protein/Peptide Name:
    CD14
  • Purity:
    >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
  • Molecular Weight:
    50 kDa, reducing conditions
  • Cat. No.:
    220024-424
  • Supplier no.:
    383-CD-050

Specifications

About this item

The Recombinant Human CD14 Protein from R&D Systems is derived from CHO. The Recombinant Human CD14 Protein has been validated for the following applications: Bioactivity.