Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Aminopeptidase P2/XPNPEP2 (from NS0 cells)
Human Recombinant Aminopeptidase P2/XPNPEP2 (from NS0 cells)
Catalog # 220023-692
Supplier:  R&D Systems
CAS Number:  
Human Recombinant Aminopeptidase P2/XPNPEP2 (from NS0 cells)
Catalog # 220023-692
Supplier:  R&D Systems
Supplier Number:  2490-ZN-010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    NS0 cells
  • Species:
    Human
  • Size:
    10 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
  • Endotoxin Content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    7512
  • Protein Synonyms:
    APP2|Membrane-bound AmP|Membrane-bound APP|EC 3.4.11.9|xaa-Pro aminopeptidase 2|XPNPEP2|Aminoacylproline aminopeptidase|mAmP|Membrane-bound aminopeptidase P|membrane-bound|X-prolyl aminopeptidase (aminopeptidase P) 2|X-Pro aminopeptidase 2
  • Accession No.:
    AAB96394
  • Protein/Peptide Name:
    Aminopeptidase P2/XPNPEP2
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Molecular Weight:
    85 kDa, reducing conditions
  • Cat. No.:
    220023-692
  • Supplier no.:
    2490-ZN-010

Specifications

About this item

The Recombinant Human Aminopeptidase P2/XPNPEP2 Protein from R&D Systems is derived from NS0. The Recombinant Human Aminopeptidase P2/XPNPEP2 Protein has been validated for the following applications: Enzyme Activity.