Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant gp130/CD130 (from Baculovirus (Sf21 Insect cells))
Human Recombinant gp130/CD130 (from Baculovirus (Sf21 Insect cells))
Catalog # 220023-541
Supplier:  R&D Systems
CAS Number:  
Human Recombinant gp130/CD130 (from Baculovirus (Sf21 Insect cells))
Catalog # 220023-541
Supplier:  R&D Systems
Supplier Number:  228-GP-050/CF
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Sf21 Insect cells)
  • Species:
    Human
  • Size:
    50 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    3572
  • Protein Synonyms:
    IL-6RB|Interleukin-6 signal transducer|CD130 antigen|IL-6 receptor subunit beta|interleukin 6 signal transducer (gp130|DKFZp564F053|IL6ST|interleukin-6 receptor subunit beta|gp130|CDw130|IL-6R subunit beta|gp130 of the rheumatoid arthritis antigenic Peptide-bearing soluble form|IL-6R-beta|CD130|oncostatin M receptor)|Oncostatin-M receptor subunit alpha|Membrane glycoprotein 130|membrane glycoprotein gp130|interleukin receptor beta chain
  • Accession No.:
    P40189
  • Protein/Peptide Name:
    gp130
  • Purity:
    >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
  • Cat. No.:
    220023-541
  • Supplier no.:
    228-GP-050/CF

Specifications

About this item

The Recombinant Human gp130 Protein from R&D Systems is derived from Sf 21 (baculovirus). The Recombinant Human gp130 Protein has been validated for the following applications: Bioactivity.