To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 μg
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
- Endotoxin Content:<1.0 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:767
- Protein Synonyms:CALScarbonate dehydratase|carbonic anhydrase-like sequence|MGC120502|CALS|CA8|carbonic anhydrase-related protein|CA-VIII|carbonic anhydrase VIIIMGC99509|CARPCA-related protein|CARP|CAMRQ3
- Accession No.:P35219
- Protein/Peptide Name:Carbonic Anhydrase VIII/CA8
- Purity:>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular Weight:38 kDa, reducing conditions
- Cat. No.:220023-463
- Supplier no.:2187-CA-050
The Recombinant Human Carbonic Anhydrase VIII Protein from R&D Systems is derived from E. coli. The Recombinant Human Carbonic Anhydrase VIII Protein has been validated for the following applications: Enzyme Activity.