To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:NS0 cells
- Species:Human
- Size:50 μg
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin Content:<0.1 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:1009
- Protein Synonyms:OB|OB-cadherin|OB-cadherin (osteoblast)|Osteoblast cadherin|cadherin 11|Cadherin11|cadherin-11|CDHOB|CDH11|CAD11OSF-4|type 2
- Accession No.:AAA35622
- Protein/Peptide Name:Cadherin-11
- Purity:>85%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular Weight:110-120 kDa, reducing conditions
- Cat. No.:220023-181
- Supplier no.:1790-CA-050
The Recombinant Human Cadherin-11 (CAD-11) Fc Chimera from R&D Systems is derived from NS0. The Recombinant Human Cadherin-11 (CAD-11) Fc Chimera has been validated for the following applications: Bioactivity.