To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:NS0 cells
- Species:Human
- Size:50 μg
- Environmentally Preferable:
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin Content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:2212
- Protein Synonyms:IgG Fc receptor II-a|Immunoglobulin G Fc receptor II|FcGR|low affinity immunoglobulin gamma Fc region receptor II-a|CD32A|receptor (CD32)|CDw32fc-gamma-RIIa|FCRIIA|FCG2|CD32MGC23887|FcRII-a|FCGR2A1|receptor for (CD32)|low affinity IIa|CD32 antigen|Fc-gamma RII-a|FCGR2|FCGR2A|IGFR2MGC30032|Fc-gamma-RIIa|Fc fragment of IgG|FcgRIIA
- Accession No.:AAA35827
- Protein/Peptide Name:Fc gamma RIIA/CD32a
- Purity:>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
- Molecular Weight:32 kDa, reducing conditions
- Cat. No.:220022-921
- Supplier no.:1330-CD-050
The Recombinant Human Fc gamma RIIA/CD32a (R167) Protein from R&D Systems is derived from NS0. The Recombinant Human Fc gamma RIIA/CD32a (R167) Protein has been validated for the following applications: Binding Activity.