Order Entry
Puerto Rico
ContactUsLinkComponent
 
Bovine FGF acidic/FGF1 (from Brain)
Catalog # 220022-920
Supplier:  R&D Systems
CAS Number:  
Bovine FGF acidic/FGF1 (from Brain)
Catalog # 220022-920
Supplier:  R&D Systems
Supplier Number:  132-FA-025/CF
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    Brain
  • Species:
    Bovine
  • Size:
    25 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    2246
  • Protein Synonyms:
    fibroblast growth factor 1 (acidic)|alpha|AFGF|endothelial cell growth factor|HBGF-1|FGF1|FGF-alpha|HBGF1|GLIO703|beta|ECGF-betaAcidic fibroblast growth factor|beta-ECGF|FGFABeta-endothelial cell growth factor|alpha-ECGF|heparin-binding growth factor 1|FGF-1|ECGF|ECGFB
  • Protein/Peptide Name:
    FGF acidic/FGF1
  • Purity:
    >97%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Molecular Weight:
    16 kDa, reducing conditions
  • Cat. No.:
    220022-920
  • Supplier no.:
    132-FA-025/CF

Specifications

About this item

The Bovine FGF acidic Protein from R&D Systems is derived from Bovine Brain. The Bovine FGF acidic Protein has been validated for the following applications: Bioactivity.