Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Mouse Recombinant Wnt-3a (from CHO cells)

Mouse Recombinant Wnt-3a (from CHO cells)

10 μg
Catalog # 220022-915
Marketsource
Supplier:   R&D Systems
Supplier #:  1324-WN-010
Mouse Recombinant Wnt-3a (from CHO cells)
Catalog # 220022-915
Supplier:  R&D Systems
Marketsource
CAS Number:  
Mouse Recombinant Wnt-3a (from CHO cells)

10 μg

CAS Number:  
Price
$1,318.89
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The Recombinant Mouse Wnt-3a Protein from R&D Systems is derived from CHO. The Recombinant Mouse Wnt-3a Protein has been validated for the following applications: Bioactivity.

Specifications

Specifications

  • Size:
    10 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Molecular Weight:
    41 kDa, reducing conditions
  • Purity:
    >75%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Endotoxin Content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Accession No.:
    P27467
  • Source:
    CHO cells
  • Cat. No.:
    220022-915
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    89780
  • Protein Synonyms:
    Wnt3a|member 3A|MGC119419|wingless-type MMTV integration site family|MGC119420|protein Wnt-3a|MGC119418
  • Protein/Peptide Name:
    Wnt-3a
  • Conjugation:
    Unconjugated
  • Species:
    Mouse

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.