Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:10 μg
- Storage Conditions:This product is stable at <= -70░ C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles.
- Gene ID:5594
- Protein Synonyms:PRKM1MAPK 2|protein tyrosine kinase ERK2|P42MAPK|Mitogen-activated protein kinase 2|PRKM2|Extracellular signal-regulated kinase 2|PRKM1|mitogen-activated protein kinase 1|MAPK2|ERK|MAP kinase 1|ERT1|p38|p41mapk|ERK-2|EC 2.7.11|MAPK 1|p40|MAPK1|p42-MAPK|ERK2MAP kinase isoform p42|EC 2.7.11.24|MAP kinase 2|p41
- Accession No.:NM_002745
- Protein/Peptide Name:ERK2
- Purity:>70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane
- Molecular Weight:68 kDa
- Cat. No.:220009-289
- Supplier no.:1230-KS-010
Specifications
About this item
The Recombinant Human Active ERK2 Protein from R&D Systems is derived from E. coli. The Recombinant Human Active ERK2 Protein has been validated for the following applications: Bioactivity.