Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant TWEAK R/TNFRSF12 (from NS0 Cells)
Human Recombinant TWEAK R/TNFRSF12 (from NS0 Cells)
Catalog # 220022-798
Supplier:  R&D Systems
CAS Number:  
Human Recombinant TWEAK R/TNFRSF12 (from NS0 Cells)
Catalog # 220022-798
Supplier:  R&D Systems
Supplier Number:  1199-TW-050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    NS0 cells
  • Species:
    Human
  • Size:
    50 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    51330
  • Protein Synonyms:
    FN14tumor necrosis factor receptor superfamily member 12A|FGF-inducible 14|CD266|Tweak-receptor|Fibroblast growth factor-inducible immediate-early response protein 14|TweakR|type I transmembrane protein Fn14|CD266 antigen|tumor necrosis factor receptor superfamily|member 12A|Fn14|TNFRSF12A|TNFRSF12
  • Accession No.:
    Q9NP84
  • Protein/Peptide Name:
    TWEAK R/TNFRSF12
  • Purity:
    >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining
  • Molecular Weight:
    35-45 kDa, reducing conditions
  • Cat. No.:
    220022-798
  • Supplier no.:
    1199-TW-050

Specifications

About this item

The Recombinant Human TWEAK R/TNFRSF12 Fc Chimera Protein from R&D Systems is derived from NS0. The Recombinant Human TWEAK R/TNFRSF12 Fc Chimera Protein has been validated for the following applications: Bioactivity.