Human IFN-alpha C/IFN-alpha 10
220009-266
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Human
- Size:100000 UN
- Storage Conditions:Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
- Gene ID:3446
- Protein Synonyms:interferon|alpha 10|IFNA10|IFNalpha C|interferon alpha-10|MGC119879|IFN-alpha-10|Interferon alpha-C|IFN-alphaC|Interferon alpha-6L|LeIF C|MGC119878
- Protein/Peptide Name:IFN-alpha C/IFN-alpha 10
- Cat. No.:220009-266
- Supplier no.:11120-1