Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Upadacitinib
Upadacitinib ≥98% (by HPLC)
Upadacitinib ≥98% (by HPLC)
Upadacitinib ≥98% (by HPLC)
Supplier:
Avantor
Certificates
Expand image
Upadacitinib ≥98% (by HPLC)
Supplier:
Avantor
About this item
CAS Number:
1310726-60-3
:
Formula:
C₁₇H₁₉F₃N₆O
MDL Number:
MFCD30502663
:
380.37 g/mol
Related Information
Documentation
Datasheet
TDS Janus Kinase JAK-1 Inhibitor, ABT-494.pdf
Documentation