pH Probe Holder and Ferrules, Ace Glass
Supplier: Ace Glass
Certificates
About this item
pH probe holder for use with Ace Glass Scale-Up Series benchtop reactor systems from 2000 to 6000 ml.
- Designed specifically for Cole-Parmer refillable general purpose probe
Unwetted section: 115L×16D mm
Wetted section: 12.3D mm
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.