Order Entry
Puerto Rico
ContactUsLinkComponent
Human Synthetic Abeta1-42 Oligomers Peptide
Human Synthetic Abeta1-42 Oligomers Peptide
Catalog # 76265-500
Supplier:  Biosensis
CAS Number:  
undefined
Human Synthetic Abeta1-42 Oligomers Peptide
Catalog # 76265-500
Supplier:  Biosensis
Supplier Number:  PE-1750-1000
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Synthetic
  • Species:
    Human
  • Size:
    1 μg
  • Storage Conditions:
    Store unopened, lyophilized oligomeric Aβ with desiccant, insulated, at –20 °C short term, –80 °C long term. Store reconstituted vial at 2- 8 °C for up to 2 days. The reconstituted material should not be frozen for best results.
  • Gene ID:
    P05067 A4_HUMAN;
  • Protease-free:
    Y
  • Protein Synonyms:
    APPI|Beta-amyloid protein 42|ABPP|Amyloid beta A4 protein|Beta-APP42
  • Protein/Peptide Name:
    Abeta1-42 oligomers Peptide
  • Molecular Weight:
    4.5 kDa (monomer); various molecular weights > 4.5 kDa for oligomeric species
  • Sequence:
    DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    ambient
  • Cat. No.:
    76265-500

Specifications

About this item

Oligomeric Human beta-Amyloid Aβ1-42 Peptide, Stabilized

A proprietary preparation of human amyloid beta peptide (amino acids 1-42) that was initially monomerized by HFIP-treatment and then allowed to form oligomers by the procedure described in Youmans KL et al., 2012, followed by lyophilisation using Biosensis’ proprietary stabilization procedures.

The resulting oligomeric mixture has been specially designed to allow the formation of stable, oligomeric Aβ1-42 peptide, multimeric complexes or oligomers. The material is intended to be used as a stable and consistent standard or positive control for oligomeric ELISA assays, as well as other research applications. This product is supplied as 2 x 500 ng vials, each containing lyophilized Aβ oligomers. Note that the amount of provided oligomeric protein is based on the amount of monomeric Aβ used to form these oligomers. The precise formation, size and number of oligomers cannot be quantified by any known method.