Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Native NGF (from Submaxillary glands)
Mouse Native NGF (from Submaxillary glands)
Supplier:  Biosensis
Certificates
undefined
Mouse Native NGF (from Submaxillary glands)
Supplier:  Biosensis
Certificates

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Neurotrophin, growth factor
  • Protein/Peptide Type:
    Native
  • Source:
    Submaxillary glands
  • Species:
    Mouse
  • Storage Conditions:
    Store lyophilized protein at 2-8C. After reconstitution, store at 2-8C short term. Store long-term at –20 °C to -80C. Avoid repeated freezing and thawing. See expiration date for shelf-life estimates, actual times may vary depending upon experimental conditions and laboratory handling.
  • Biological Activity:
    Tested by stimulation of 8 day old chick embryo dorsal root ganglia or neurite outgrowth in rat PC12 cells.
  • Gene ID:
    P01139 NGF_MOUSE
  • Reconstitution Instructions:
    Reconstitute with the addition of 500 µL sterile water and divide into single use aliquots. Concentration after reconstitution: 1 mg/mL
  • Protein Synonyms:
    beta NGF|2.5S NGF
  • Protein/Peptide Name:
    NGF
  • Purity:
    Greater than 90% (as determined by SDS electrophoresis)
  • Molecular Weight:
    The homodimer has a molecular weight of ~26KDa and the monomer ~13KDa as confirmed by SDS PAGE.
  • Formulation:
    Lyophilised from PBS, pH 7.4 without preservatives.
  • Shipping Temperature:
    ambient

Specifications

Related Information