Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Black Gold II Myelin Staining Kit
Black Gold II Myelin Staining Kit
Black Gold II Myelin Staining Kit
Black Gold II Myelin Staining Kit
Catalog # 76264-654
Supplier:
Biosensis
CAS Number:
Expand image
Black Gold II Myelin Staining Kit
Catalog # 76264-654
Supplier:
Biosensis
Supplier Number:
TR-100-BG
CAS Number:
Specifications
Size:
1 kit
Cat. No.:
76264-654
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Ambient
Product Details
Documentation