Anti-PF-4 (CXCL4) Rabbit Polyclonal Antibody (Biotin)
Produced from sera of rabbits immunized with highly pure Recombinant Human PF-4 (CXCL4). AntiHuman PF-4 (CXCL4)specific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: PF-4 (CXCL4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 10775-000
- 10775-002
- Antigen Symbol:
- PF-4 (CXCL4)
- PF-4 (CXCL4)
- Antigen Name:
- PF-4
- PF-4
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human PF-4 (CXCL4)
- Recombinant Human PF-4 (CXCL4)
- Form:
- Lyophilized
- Lyophilized
- Molecular Weight:
- 7.8 kDa
- 7.8 kDa
- Purification:
- Recombinant Human PF-4 (CXCL4)
- Recombinant Human PF-4 (CXCL4)
- Storage Buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage Temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient
Frequently Bought Together