Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MCP-1 (CCL2) Rabbit Polyclonal Antibody (Biotin)
Anti-MCP-1 (CCL2) Rabbit Polyclonal Antibody (Biotin)
Supplier:  Peprotech
Certificates
Anti-MCP-1 (CCL2) Rabbit Polyclonal Antibody (Biotin)
Supplier:  Peprotech

Specifications

  • Catalog No:
  • 10775-250
  • 10775-248
  • Antigen Symbol:
  • MCP-1 (CCL2)
  • MCP-1 (CCL2)
  • Antigen Name:
  • MCP-1
  • MCP-1
  • Antibody Type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human MCP-1 (CCL2)
  • Recombinant Human MCP-1 (CCL2)
  • Form:
  • Lyophilized
  • Lyophilized
  • Molecular Weight:
  • 8.6 kDa
  • 8.6 kDa
  • Purification:
  • Recombinant Human MCP-1 (CCL2)
  • Recombinant Human MCP-1 (CCL2)
  • Storage Buffer:
  • See Product Data Sheet for more information.
  • See Product Data Sheet for more information.
  • Storage Temperature:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping Temperature:
  • Ambient
  • Ambient

Specifications

Frequently Bought Together