Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-MCP-4 (CCL13) Rabbit Polyclonal Antibody
 :  Peprotech
 
Anti-MCP-4 (CCL13) Rabbit Polyclonal Antibody
 :  Peprotech

 

  • Catalog No:
  • 10774-970
  • 10774-974
  • 10774-972
  • Antigen Symbol:
  • MCP-4 (CCL13)
  • MCP-4 (CCL13)
  • MCP-4 (CCL13)
  • Antigen Name:
  • MCP-4
  • MCP-4
  • MCP-4
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human MCP-4 (CCL13)
  • Recombinant Human MCP-4 (CCL13)
  • Recombinant Human MCP-4 (CCL13)
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Molecular Weight:
  • 8.6 kDa
  • 8.6 kDa
  • 8.6 kDa
  • Purification:
  • Recombinant Human MCP-4 (CCL13)
  • Recombinant Human MCP-4 (CCL13)
  • Recombinant Human MCP-4 (CCL13)
  • Storage Buffer:
  • See Product Data Sheet for more information.
  • See Product Data Sheet for more information.
  • See Product Data Sheet for more information.
  • Storage Temperature:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping Temperature:
  • Ambient
  • Ambient
  • Ambient

 

Frequently Bought Together