Anti-MCP-1 (CCL2) Rabbit Polyclonal Antibody
Supplier: Peprotech
Certificates
About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human MCP-1 (CCL2). AntiHuman MCP-1 (CCL2)specific antibody was purified by affinity chromatography employing an immobilized Human MCP-1 matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: MCP-1 (CCL2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 10775-246
- 10775-242
- 10775-244
- Antigen Symbol:
- MCP-1 (CCL2)
- MCP-1 (CCL2)
- MCP-1 (CCL2)
- Antigen Name:
- MCP-1
- MCP-1
- MCP-1
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant Human MCP-1 (CCL2)
- Recombinant Human MCP-1 (CCL2)
- Recombinant Human MCP-1 (CCL2)
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular Weight:
- 8.6 kDa
- 8.6 kDa
- 8.6 kDa
- Purification:
- Recombinant Human MCP-1 (CCL2)
- Recombinant Human MCP-1 (CCL2)
- Recombinant Human MCP-1 (CCL2)
- Storage Buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage Temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient
- Ambient
Specifications
Frequently Bought Together