Anti-IGF-BP5 Rabbit Polyclonal Antibody
Catalog # 10777-078
Supplier: Peprotech
Specifications
- Antibody Type:Primary
- Antigen Name:IGF-BP5
- Antigen Symbol:IGF-BP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:1 mg
- Form:Lyophilized
- Storage Buffer:See Product Data Sheet for more information.
- Molecular Weight:28.6 kDa
- Storage Temperature:Store at –20 °C prior to reconstitution.
- Shipping Temperature:Ambient
- Immunogen:Recombinant Human IGF-BP5
- Purification:Recombinant Human IGF-BP5
- Cat. No.:10777-078
- Packaging:PP Vial
- Supplier no.:500P2321MG
Specifications
About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human IGF-BP5. AntiHuman IGF-BP5specific antibody was purified by affinity chromatography employing an immobilized Human IGF-BP5 matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: IGF-BP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human