Anti-gAcrp30/Adipolean Variant Rabbit Polyclonal Antibody
Supplier: Peprotech
Certificates
About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human gAcrp30/Adipolean Variant. AntiHuman gAcrp30/Adipolean Variantspecific antibody was purified by affinity chromatography employing an immobilized Human gAcrp30/Adipolean Variant matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: gAcrp30/Adipolean Variant
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 10776-870
- 10776-866
- 10776-868
- Antigen Symbol:
- gAcrp30/Adipolean Variant
- gAcrp30/Adipolean Variant
- gAcrp30/Adipolean Variant
- Antigen Name:
- gAcrp30/Adipolean
- gAcrp30/Adipolean
- gAcrp30/Adipolean
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant Human gAcrp30/Adipolean Variant
- Recombinant Human gAcrp30/Adipolean Variant
- Recombinant Human gAcrp30/Adipolean Variant
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular Weight:
- 18.1 kDa
- 18.1 kDa
- 18.1 kDa
- Purification:
- Recombinant Human gAcrp30/Adipolean Variant
- Recombinant Human gAcrp30/Adipolean Variant
- Recombinant Human gAcrp30/Adipolean Variant
- Storage Buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage Temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient
- Ambient
Specifications
Frequently Bought Together