Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant sRANK (from CHO cells)
Mouse Recombinant sRANK (from CHO cells)
Supplier:  Peprotech
Certificates
Mouse Recombinant sRANK (from CHO cells)
Supplier:  Peprotech

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Storage Conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin Content:
    >0.1 ng/μg
  • Biological Activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Reconstitution Instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein Synonyms:
    TRANCE (TNF-Related Activation-induced Cytokine)|OPGL|TNFSF11|CD254|ODF (Osteoclast Differentiation Factor)|soluble Receptor Activator of NF-κB Ligand
  • Protein/Peptide Name:
    sRANK
  • Molecular Weight:
    kDa
  • Sequence:
    FSGAPAMMEG SWLDVAQRGK PEAQPFAHLT INAASIPSGS HKVTLSSWYH DRGWAKISNM TLSNGKLRVN QDGFYYLYAN ICFRHHETSG SVPTDYLQLM VYVVKTSIKI PSSHNLMKGG STKNWSGNSE FHFYSINVGG FFKLRAGEEI SIQVSNPSLL DPDQDATYFG AFKVQDID
  • Endotoxin Level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping Temperature:
    Ambient

Specifications