Order Entry
Puerto Rico
ContactUsLinkComponent
Human;Mouse;Rat Recombinant Irisin (from CHO cells)
Human;Mouse;Rat Recombinant Irisin (from CHO cells)
Supplier:  Peprotech
Certificates
Human;Mouse;Rat Recombinant Irisin (from CHO cells)
Supplier:  Peprotech

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
    Rat
    Mouse
  • Storage Conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin Content:
    >0.1 ng/μg
  • Biological Activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Gene ID:
    252995 Q8NAU1
  • Reconstitution Instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein Synonyms:
    Fibronectin type III domain-containing protein 5 (FNDC5)|Fibronectin type III repeat-containing protein 2 (FRCP2)
  • Protein/Peptide Name:
    Irisin
  • Purity:
    0.9
  • Molecular Weight:
    25.2 kDa
  • Sequence:
    DSPSAPVNVT VRHLKANSAV VSWDVLEDEV VIGFAISQQK KDVRMLRFIQ EVNTTTRSCA LWDLEEDTEY IVHVQAISIQ GQSPASEPVL FKTPREAEKM ASKNKDEVTM KE
  • Endotoxin Level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping Temperature:
    Ambient

Specifications

Frequently Bought Together